Learn More
Cayman Chemical ANTIBODY BAR502 1mg
SDP
Supplier:  Cayman Chemical 224621 MG
Encompass_Preferred
A dual agonist of GP-BAR1 and FXR (EC50s = 0.2 and 1 μM, respectively); increases the expression of FXR-regulated BSEP, OSTα, and SHP in primary hepatocytes isolated from FXR wild-type mice; increases cAMP-luciferase reporter gene expression in HEK293T cells
Catalog No. 50-298-7353