Learn More
Cayman Chemical ANTIBODY BAY-299 25mg
SDP
Supplier:  Cayman Chemical 1977725 MG
Encompass_Preferred
A potent and selective BRD1 inhibitor (IC50 = 6 nM); also inhibits the second bromodomain of TAF1 (IC50 = 13 nM); >30-fold selective over BRPF1, BRPF3, BRD9, and ATAD2; >300-fold selective over BRD4
Catalog No. 50-298-5419