Learn More
Cayman Chemical ANTIBODY BB-F-Yn 500ug
SDP
Supplier:  Cayman Chemical 25379500 UG
Encompass_Preferred
A clickable PAD inhibitor; inhibits PAD1-4 (kinact/KI values of 900, 1,200, 3,400, and 3,750 M-1min-1, respectively); has been used for labeling PADs in cell-free and cell-based assays, followed by click reactions with azide-modified TAMRA reporters or biotin capture reagents
Catalog No. 50-293-2727