Learn More
Cayman Chemical ANTIBODY Bendazac 1g
SDP
Supplier:  Cayman Chemical 351271 G
Encompass_Preferred
An NSAID; prevents denaturation of lens and serum proteins in vitro; prevents cyanate-induced opacification of the eye lens in a rat model of cataract from 10-40 mM
Catalog No. 50-291-1067