Learn More
Cayman Chemical ANTIBODY BH3I-1 5mg
SDP
Supplier:  Cayman Chemical 187635 MG
Encompass_Preferred
A cell permeable inhibitor that blocks the binding of BH3 peptides to Bcl-xL, inducing apoptosis; inhibits interactions of BH3 domain-containing proteins with Bcl-xL, Bcl-2, and Bcl-W, inducing apoptosis in Bcl-2 or Bcl-W expressing cells with Ki values of 43.4 and 124 µM, respectively
Catalog No. 50-298-0319