Learn More
Cayman Chemical ANTIBODY BI-3406 5mg
SDP
Supplier:  Cayman Chemical 352615 MG
Encompass_Preferred
An inhibitor of the K-Ras-SOS1 protein-protein interaction (IC50 = 5 nM); selectively disrupts the protein-protein interaction between K-RasG12C or K-RasG12D with SOS1 over SOS2, and it is selective over 321 kinases in a panel at 10 µM but does inhibit PLK4, BMPR2, and LRRK2 activity by >50%
Catalog No. 50-291-3542