Learn More
Cayman Chemical ANTIBODY BIapenm 10mg
SDP
Supplier:  Cayman Chemical 2882210 MG
Encompass_Preferred
A broad-spectrum β-lactam antibiotic; active against aerobic and anaerobic, Gram-positive and Gram-negative bacteria, including clinical isolates of E. coli, oxacillin-sensitive S. aureus, B. fragilis, and Clostridium (MIC90s = 0.12, 0.12, 0.25, and 1 μg/ml, respectively); active against the rifampicin-susceptible H37Rv and rifampicin-resistant 115R and 124R M. tuberculosis strains (MICs = 2-4, 4-8, and 8-16 mg/L, respectively); decreases the number of lung CFUs in a mouse model of M. tuberculosis infection at 200 mg/kg
Catalog No. 50-293-8184