Learn More
Cayman Chemical ANTIBODY BIIB-057 10mg
SDP
Supplier:  Cayman Chemical 1765210 MG
Encompass_Preferred
A potent, selective, orally bioavailable inhibitor of Syk (IC50 = 1 nM); blocks B cell receptor-mediated cell signaling and activation in whole blood (IC50s = 0.27 and 0.28 µM, respectively); inhibits inflammation and tumor growth in animal models of rheumatoid arthritis and non-Hodgkin lymphoma, respectively
Catalog No. 50-296-6274