Learn More
Cayman Chemical ANTIBODY BIkInIn 25mg
SDP
Supplier:  Cayman Chemical 1838125 MG
Encompass_Preferred
An inhibitor of several ASK isoforms when given at a concentration of 10 µM, leaving residual kinase activity at less than 10% for ASKα, γ, ε, ζ, η, ι, and 20% for ASKθ, without inhibiting isoforms β and δ; activates signaling induced by brassinosteroids
Catalog No. 50-298-0056