Learn More
Cayman Chemical ANTIBODY BItertnol 500mg
SDP
Supplier:  Cayman Chemical 24054500 MG
Encompass_Preferred
A triazole fungicide; active against a variety of fungi, including isolates of V. inaequalis (MICs = 0.6 and 1 UG/ml) but repeated use leads to resistance (MICs = 9.8-13 UG/ml for resistant isolates); inhibits CYP3A4 (IC50 = 2.74 µM) and DHT-induced androgenic activity (IC50 = 79.85 µM); increases operant responding in rats (10-300 mg/kg, i.p.)
Catalog No. 50-293-3261