Learn More
Cayman Chemical ANTIBODY BIX02189 10mg
SDP
Supplier:  Cayman Chemical 1699910 MG
Encompass_Preferred
A potent inhibitor of both MEK5 and ERK5 (IC50s = 1.5 and 59 nM, respectively); minimally inhibits a panel of related kinases, including MEK1/2 and ERK1/2; blocks sorbitol-induced phosphorylation of ERK5, without affecting phosphorylation of ERK1/2, in HeLa cells
Catalog No. 50-296-5395