Learn More
Cayman Chemical ANTIBODY BLU-285 25mg
SDP
Supplier:  Cayman Chemical 2298525 MG
Encompass_Preferred
A dual inhibitor of KIT and PDGFRα mutant kinases (IC50s = 0.27 and 0.24 nM for KITD816V and PDGFRαD842V, respectively); >150-fold selective for KITD816V and PDGFRαD842V over a kinase panel at a concentration of 3 μM; has activity against a panel of KIT and PDGFRα loop mutants identified in patients with GISTs (IC50s = D814Y mastocytoma allograft mouse model and a GIST patient-derived mouse xenograft model in a dose-dependent manner when administered at doses ranging from 0.3-30 mg/kg
Catalog No. 50-299-4997