Learn More
Cayman Chemical ANTIBODY BLU-554 5mg
SDP
Supplier:  Cayman Chemical 224385 MG
Encompass_Preferred
A potent inhibitor of recombinant FGFR4 (IC50 50 < 10 nM), a marker of FGFR4 inhibition, in MDA-MB453 cells
Catalog No. 50-298-7354