Learn More
Cayman Chemical ANTIBODY BLX3887 1mg
SDP
Supplier:  Cayman Chemical 273911 MG
Encompass_Preferred
A 15-LO-1 inhibitor (IC50 = 32 nM); selective for 15-LO-1 over 15-LO-2, which it does not inhibit, 5-LO (IC50 = 472 nM) and 12-LO (IC50 = 3,310 nM); inhibits endocytosis in, and the migration of, isolated human peripheral blood mononuclear cell-derived dendritic cells in vitro
Catalog No. 50-293-7054