Learn More
Cayman Chemical ANTIBODY BMS 191011 5mg
SDP
Supplier:  Cayman Chemical 222995 MG
Encompass_Preferred
A KCa1.1/BK channel activator; increases maximum potassium current to 126% of control X. laevis oocytes expressing human KCa1.1 channels at 1 μM; increases the diameter of retinal arterioles in rats from 10-100 μg/kg
Catalog No. 50-298-7676