Learn More
Cayman Chemical ANTIBODY BMS 466442 1mg
SDP
Supplier:  Cayman Chemical 420751 MG
Encompass_Preferred
An Asc-1 inhibitor; selectively inhibits Asc-1 in HEK293 cells expressing Asc-1 (IC50 = 36.8 nM) over 40 other transporters (IC50s = >10 µM); inhibits serine, cysteine, glycine, and alanine import and cAMP-induced increases in the OCR and decreases UCP1 levels in primary human white and brown adipose cells at 100 nM; decreases the number of malformed neurons and reduces contextual fear memory deficits in a mouse model of surgery-induced brain damage when intracerebroventricularly infused
Catalog No. 50-294-5150