Learn More
Cayman Chemical ANTIBODY BohemIn 5mg
SDP
Supplier:  Cayman Chemical 210735 MG
Encompass_Preferred
A Cdk inhibitor (IC50s = 4.6, 8.3, and 2.7 µM for Cdk2/cyclin E, Cdk2/cyclin A, Cdk9/cyclin T1, respectively); suppresses tumor cell growth, inducing G1 and G2 phase cell cycle arrest
Catalog No. 50-298-1161