Learn More
Cayman Chemical ANTIBODY BPR1J-097 50mg
SDP
Supplier:  Cayman Chemical 2980550 MG
Encompass_Preferred
An FLT3 inhibitor (IC50 = 11 nM); selective for FLT3 over FLT1, KDR, Aurora A, and Aurora B kinases (IC50s = 211, 129, 340, and 876 nM, respectively); inhibits FLT3 activity and phosphorylation of STAT5 in MV4-11 AML cells containing the FLT3 activating mutant FLT3-ITD (IC50s = 10 and 1 nM, respectively); inhibits the growth of FLT3-dependent MOLM-13 and MV4-11 AML cells (GI50s = 21 and 46 nM, respectively); inhibits tumor growth in a MOLM-13 mouse xenograft model at 25 mg/kg
Catalog No. 50-298-9719