Learn More
Cayman Chemical ANTIBODY BrazIlIn 10mg
SDP
Supplier:  Cayman Chemical 2245610 MG
Encompass_Preferred
A flavonoid with diverse bioactivities; inhibits Aβ (1-42) fibrillogenesis (IC50 = 1.5 µM); prevents Aβ (1-42) mature fibril remodeling; inhibits the production of cytokines, including PGE2 and TNF-α (IC50s = 12.6 and 87.2 µM); effective against Gram-positive and Gram-negative bacteria MICs = 31.3-250 UG/ml; inhibits RANKL-mediated osteoclast differentiation; protective against LPS-induced osteoporosis in mice (100 mg/kg)
Catalog No. 50-298-6955