Learn More
Cayman Chemical ANTIBODY BRD7389 10mg
SDP
Supplier:  Cayman Chemical 2021410 MG
Encompass_Preferred
An inhibitor of RSKs (IC50s = 1.5, 2.4, and 1.2 μM for RSK1, RSK2, and RSK3, respectively); causes mouse αTC1 pancreatic α cells to adopt several morphological and gene expression features of β cells, including increased insulin expression
Catalog No. 50-298-5417