Learn More
Cayman Chemical ANTIBODY BRD73954 10mg
SDP
Supplier:  Cayman Chemical 1691910 MG
Encompass_Preferred
A dual inhibitor of HDAC6 and HDAC8 (IC50s = 36 and 120 nM, respectively); selective for HDAC6 and -8 over HDAC1-5, -7, and -9 (IC50s = 12, 9, 23, >33, >33, 13, and >33 µM, respectively); increases acetylation of α-tubulin in HeLa cells at 10 µM
Catalog No. 50-296-6945