Learn More
Cayman Chemical ANTIBODY BRD7552 25mg
SDP
Supplier:  Cayman Chemical 1914325 MG
Encompass_Preferred
An inducer of PDX1 expression; increases PDX1 and insulin gene expression in PANC-1 cells; increases PDX1 protein levels in PANC-1 cells; increases histone H3 acetylation and trimethylation of H3K4 and H3K9
Catalog No. 50-298-3643