Learn More
Cayman Chemical ANTIBODY BudesonIde 100mg
SDP
Supplier:  Cayman Chemical 15407100 MG
Encompass_Preferred
A glucocorticoid and an agonist of glucocorticoid receptors (EC50 = 45.7 pM in a transactivation assay); selective for glucocorticoid over mineralocorticoid receptors (EC50 = 7,620 pM); inhibits LPS-induced TNF-α release from human PBMCs (IC500 = 0.96 nM); reduces levels of IL-1β and eotaxin in the lungs and the number of eosinophils and neutrophils in BALF in a rat model of ovalbumin-induced airway inflammation at 3 mg/kg; intracolonic administration decreases colon wet weight and colonic MPO activity in a rat model of oxazolone-induced colitis
Catalog No. 50-296-8534