Learn More
Cayman Chemical ANTIBODY Butafosfn 5g
SDP
Supplier:  Cayman Chemical 186135 G
Encompass_Preferred
An organic phosphorus supplement that is given, most commonly with cyanocobalamin, to cattle, swine, horses, and poultry for the prevention or treatment of deficiencies
Catalog No. 50-296-6160