Learn More
Cayman Chemical ANTIBODY ButeIn 1mg
SDP
Supplier:  Cayman Chemical 153711 MG
Encompass_Preferred
A plant polyphenol that is classified as a chalcone; a potent antioxidant that affects the numerous pathways modulated by oxidant tone, including inflammation, cancer, and immune response
Catalog No. 50-296-9432