Learn More
Cayman Chemical ANTIBODY BVT 14225 1mg
SDP
Supplier:  Cayman Chemical 412241 MG
Encompass_Preferred
An 11β-HSD1 inhibitor (IC50s = 52 and 284 for the human and mouse enzymes, respectively); selective for human 11β-HSD1 over human 11β-HSD2 (IC50 = >10,000 nM); inhibits 11β-HSD1 by 55% in HEK293 cells expressing human HSD11B1 at 10 µM; reduces blood glucose levels in a rat model of streptozotocin- and nicotinamide-induced diabetes at 50 mg/kg
Catalog No. 50-294-3153