Learn More
Cayman Chemical ANTIBODY BZAD-01 5mg
SDP
Supplier:  Cayman Chemical 387885 MG
Encompass_Preferred
An antagonist of NR2B subunit-containing NMDA receptors (Ki = 72 nM); selective for NR2B subunit-containing NMDA receptors over those containing NR2A subunits (IC50s = 47 and >10,000 nM, respectively); reduces carrageenan-induced hyperalgesia in rats (ED50 = 5.5 mg/kg); reduces dopaminergic neuron loss in the rat substantia nigra pars compacta and improves measures of ipsilateral curling, ipsilateral head positioning, and beam walking in a rat model of 6-OHDA-induced Parkinson’s disease at 10 mg/kg
Catalog No. 50-291-7904