Learn More
Cayman Chemical ANTIBODY C-176 25mg
SDP
Supplier:  Cayman Chemical 2585925 MG
Encompass_Preferred
A STING inhibitor; reduces STING-, but not RIG-I or TBK1-, mediated IFN-β reporter activity in HEK293 cells from 0.01-5 μM; reduces serum levels of type-I IFNs and cardiac inflammation the in Trex-/- mouse model of multi-organ inflammation
Catalog No. 50-293-6804