Learn More
Cayman Chemical ANTIBODY C-6 NBD 1g
SDP
Supplier:  Cayman Chemical 188711 G
Encompass_Preferred
A derivative of NBD modified with a hexanoic acid tail, which provides a reactive carboxylic acid site; used to synthesize a variety of fluorescently tagged compounds for research purposes
Catalog No. 50-298-1414