Learn More
Cayman Chemical ANTIBODY C-87 5mg
SDP
Supplier:  Cayman Chemical 365785 MG
Encompass_Preferred
An inhibitor of TNF-α; binds to TNF-α; inhibits TNF-α-induced cytotoxicity (IC50 = 8.73 µM) in L-929 cells; reduces liver inflammation and injury, as well as increases survival, in a mouse model of LPS- and GalN-induced acute hepatitis at 12.5 mg/kg
Catalog No. 50-290-9493