Learn More
Cayman Chemical ANTIBODY Calhex 231 5mg
SDP
Supplier:  Cayman Chemical 172685 MG
Encompass_Preferred
A negative allosteric modulator of CaSR that blocks calcium-mediated activation (IC50 = 0.39 µM); attenuates the activation of CaSR by calcimimetics
Catalog No. 50-296-7655