Learn More
Cayman Chemical ANTIBODY CALX8 250mg
SDP
Supplier:  Cayman Chemical 24777250 MG
Encompass_Preferred
A calixarene detergent; has been used to solubilize membrane proteins; CMC = 2.2 mM
Catalog No. 50-301-5973