Learn More
Cayman Chemical ANTIBODY Carbadox 25g
SDP
Supplier:  Cayman Chemical 2490325 G
Encompass_Preferred
An inhibitor of FAF1 inhibits chemical hypoxia-induced apoptosis in H9c2 cardiomyocytes and hydrogen peroxide-induced cell death in SH-SY5Y cells at 10 and 20 M respectively reduces decreases in striatal dopamine transporter activity in a mouse model of MPTP-induced Parkinsons disease at 20 mg/kg per day
Catalog No. 50-301-6014