Learn More
Cayman Chemical ANTIBODY Carbazern 5mg
SDP
Supplier:  Cayman Chemical 260925 MG
Encompass_Preferred
An inhibitor of PDE2 and PDE3; inhibits cAMP hydrolysis (IC50 = 4.1 µM in rabbit heart ventricles) selectively over cGMP hydrolysis (IC50 = 171 µM); induces positive inotropic (EC50 = 100 µM) and negative chronotropic effects in isolated rabbit papillary muscle and right atria, respectively; a substrate for AOX1
Catalog No. 50-293-6264