Learn More
Cayman Chemical ANTIBODY CarIporIde 100mg
SDP
Supplier:  Cayman Chemical 16935100 MG
Encompass_Preferred
An NHE-1 inhibitor (IC50 = 0.56 µM in an intracellular pH recovery assay); selective for NHE-1 over NHE-2 (IC50 = 28 µM in an intracellular pH recovery assay); inhibits sodium propionate-induced swelling of isolated human platelets (IC50 = 0.023 µM); reduces the myocardial infarct size as a percentage of the area at risk in a rabbit model of ischemia-reperfusion injury induced by coronary artery occlusion at 1 mg/kg; decreases ventricular tachycardia duration in a rat model of coronary artery ligation and reperfusion-induced arrhythmia at 1 mg/kg
Catalog No. 50-296-5683