Learn More
Cayman Chemical ANTIBODY CAY10580 5mg
SDP
Supplier:  Cayman Chemical 168355 MG
Encompass_Preferred
An 8-aza-9-oxo-15-hydroxy saturated analog of PGE2; selectively binds the EP4 receptor (Ki = 35 nM) relative to the EP1, EP2, and EP3 receptors (Ki = 3,000, 2,000, and >3,000 nM, respectively); stimulates cAMP formation in excised mouse ovaries
Catalog No. 50-298-0492