Learn More
Cayman Chemical ANTIBODY CAY10581 5mg
SDP
Supplier:  Cayman Chemical 168385 MG
Encompass_Preferred
A naphthoquinone derivative that potently and reversibly inhibits indoleamine 2,3-dioxygenase (IC50 = 55 nM); a more potent inhibitor of IDO than annulin B or 1-methyl-d-tryptophan (1MT)
Catalog No. 50-297-9829