Learn More
Cayman Chemical ANTIBODY CAY10614 10mg
SDP
Supplier:  Cayman Chemical 1361510 MG
Encompass_Preferred
An antagonist of lipid A activation of TLR4 (IC50 =1.68 μM), in a cell-based assay; significantly improves survival of mice given intraperitoneal LPS
Catalog No. 50-295-5397