Learn More
Cayman Chemical ANTIBODY CAY10700 5mg
SDP
Supplier:  Cayman Chemical 189055 MG
Encompass_Preferred
An orally bioavailable inhibitor of mPGES-1 (IC50 = 0.24 µM in an in vitro human whole blood assay); inactive against COX-1 and COX-2 (9% inhibition at 100 µM and 16% at 30 µM, respectively)
Catalog No. 50-298-2196