Learn More
Cayman Chemical ANTIBODY CAY10704 10mg
SDP
Supplier:  Cayman Chemical 1924010 MG
Encompass_Preferred
A potent inhibitor of HCV infection (EC50 = 17 nM) that displays low cytotoxicity of virally-infected human hepatoma Huh7.5.1 cells (CC50 = 21.3 µM); displays good pharmacokinetics in mice when delivered intraperitoneally, with preferential liver distribution without significant hepatotoxicity
Catalog No. 50-298-4612