Learn More
Cayman Chemical ANTIBODY CAY10753 1mg
SDP
Supplier:  Cayman Chemical 302771 MG
Encompass_Preferred
A TNKS2 inhibitor (IC50 = 0.3 nM); selective for TNKS2 over TNKS1 (IC50 = 6.1 nM), as well as PARP1 and PARP2 (IC50s = 89.6 and 37.8 nM, respectively); inhibits proliferation of DLD-1 cells at 1 µM
Catalog No. 50-299-2114