Learn More
Cayman Chemical ANTIBODY CAY10762 10mg
SDP
Supplier:  Cayman Chemical 3057610 MG
Encompass_Preferred
An inhibitor of MAGL (IC50 = 34.1 nM); reduces hydrogen peroxide-induced LDH release from Neuro2a cells at 1 µM; increases levels of 2-AG in mouse brain at 10 mg/kg
Catalog No. 50-299-0653