Learn More
Cayman Chemical ANTIBODY CAY10771 5mg
SDP
Supplier:  Cayman Chemical 317585 MG
Encompass_Preferred
A dual agonist of FXR and PPARδ; activates FXR and PPARδ in reporter assays using HEK293T cells (EC50s = 0.94 and 1.5 µM, respectively); selective for these receptors over RARα, RXRα, PPARα, PPARγ, and LXRα at 10 µM
Catalog No. 50-299-0706