Learn More
Cayman Chemical ANTIBODY CBGAQ 5mg
SDP
Supplier:  Cayman Chemical 317725 MG
Encompass_Preferred
An analytical reference standard categorized as an oxidative quinone product of CBGA; intended for research and forensic applications
Catalog No. 50-299-1169