Learn More
Cayman Chemical ANTIBODY CBGH 5mg
SDP
Supplier:  Cayman Chemical 333965 MG
Encompass_Preferred
An analytical reference standard categorized as a hexyl-chain homolog of CBG; intended for research and forensic applications
Catalog No. 50-294-7312