Learn More
Cayman Chemical ANTIBODY CC-401 1mg
SDP
Supplier:  Cayman Chemical 188671 MG
Encompass_Preferred
A potent, selective inhibitor of all three JNK isoforms (Ki values range from 25 to 50 nM); bioavailable when delivered by gavage, blocking JNK signaling and renal fibrosis in a rat obstructed kidney model; decreases hepatic necrosis and apoptosis after orthotopic liver transplantation in rats
Catalog No. 50-298-1044