Learn More
Cayman Chemical ANTIBODY CC-671 1mg
SDP
Supplier:  Cayman Chemical 261841 MG
Encompass_Preferred
A dual inhibitor of Mps1/TTK and Clk2 (IC50s = 5 and 3 nM, respectively); selective for Mps1/TTK and Clk2 over a panel of 255 kinases at 3 μM; inhibit DYRK3, DYRK1A, PHKG, DYRK1B, and Clk1 (IC50s = 99, 104, 136, 157, and 300 nM, respectively); selectively inhibits the growth of Cal-51 TNBC cell over BT-474 luminal breast cancer cells (IC50s = 60 and 6,970 nM, respectively); reduces tumor volume in a Cal-51 mouse xenograft model at 20 mg/kg
Catalog No. 50-293-6879