Learn More
Cayman Chemical ANTIBODY CCT007093 25mg
SDP
Supplier:  Cayman Chemical 1999225 MG
Encompass_Preferred
A PP1D inhibitor (IC50 = 8.4 µM); selectively reduces colony formation in MCF-7 and MCF-3B cells that overexpress PP1D over HeLa cells that express wild-type levels of PPD1D (IC50s = 0.48, 0.13, and >50 µM, respectively); active against T. cruzi epimastigotes (IC50 = 6.5 µM)
Catalog No. 50-298-0671