Learn More
Cayman Chemical ANTIBODY CCT251236 1mg
SDP
Supplier:  Cayman Chemical 277881 MG
Encompass_Preferred
An inhibitor of Hsf1-mediated transcription; binds to Pirin (Ki = 28 nM) and inhibits Hsf1-mediated induction of Hsp27 in U2OS cells (IC50 = 19 nM); inhibits growth of SKOV3 ovarian carcinoma cells (GI50 = 1.1 nM); reduces tumor volume in an SKOV3 mouse xenograft model at 20 mg/kg
Catalog No. 50-293-8540