Learn More
Cayman Chemical ANTIBODY CD73-IN-1 500ug
SDP
Supplier:  Cayman Chemical 30959500 UG
Encompass_Preferred
An inhibitor of CD73 (IC50 = ≤316.23 nM for the human FLAG-tagged enzyme in a cell-free assay)
Catalog No. 50-298-9603